Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FGF-8a Recombinant Protein, PeproTech®

Catalog No. 10025A01M
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-025-A01M 2 x 500 μg
10025A100UG 100 μg
10025A250UG 250 μg
10025A25UG 25 μg
10025A500UG 500 μg
100-25A-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 10-025-A01M Supplier Gibco™ Supplier No. 10025A01M
Add to Cart
Add to Cart

Recombinant Protein

100-00CC-500UG will be provided as 2 x 250 μg (100-00CC-250UG). CHO cell-derived Recombinant Human FGF-8a is a 21.2 kDa protein consisting of 182 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF-8 is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8a, FGF-8b, FGF-8e and FGF-8f. The human and murine FGF-8a and FGF-8b are identical, unlike human and mouse FGF-8e and FGF-8f, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P55075
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 2253
Molecular Weight (g/mol) 21.2 kDa
Name Human FGF-8 A
Quantity 2 x 500 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias ace; cb110; etID309886.13; fb73a06; fgf8; fgf-8; ibd5031
Common Name FGF-8 A
Gene Symbol FGF8
Biological Activity Determined by a cell proliferation assay using balb/c 3T3 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence QHVREQSLVT DQLSRRLIRT YQLYSRTSGK HVQVLANKRI NAMAEDGDPF AKLIVETDTF GSRVRVRGAE TGLYICMNKK GKLIAKSNGK GKDCVFTEIV LENNYTALQN AKYEGWYMAF TRKGRPRKGS KTRQHQREVH FMKRLPRGHH TTEQSLRFEF LNYPPFTRSL RGSQRTWAPE PR
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.