Learn More
FUJIFILM IRVINE SCIENTIFIC INC Human FGF acidic (FGF-1), Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 1002910UG
Encompass_Preferred
Acidic fibroblast growth factor (FGF-acidic), also known as FGF-1, is a potent inducer of DNA synthesis, cell proliferation, and has chemotactic activities.  FGF-acidic regulates cardiogenesis through protein kinase C signaling.  FGF-acidic also functions as an insulin sensitizer and mediates adipose tissue remodeling.  High serum levels of FGF-acidic are associated with type 2 diabetes mellitus (T2DM), suggesting a pathogenic role of FGF-acidic during T2DM. 
Catalog No. 50-990-737
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.