Learn More
Invitrogen™ Human FGF-basic (FGF-2/bFGF) (147 aa) Recombinant Protein
Description
Fibroblast Growth Factor basic (human) Recombinant Protein, RP-8627, from Invitrogen.
Specifications
Specifications
| Accession Number | P09038 |
| Concentration | 0.1mg/mL |
| For Use With (Application) | Control |
| Formulation | 10mM sodium phosphate with 75mM NaCl and no preservative; pH 7.5 |
| Gene ID (Entrez) | 2247 |
| Molecular Weight (g/mol) | ∽16.5 |
| Name | Human FGF basic |
| pH Range | 7.5 |
| Quantity | 50 μg |
| Source | E. coli |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.