Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human FGF basic/FGF2/bFGF Magnetic Luminex™ Performance Assay
SDP

Catalog No. LUHM233 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LUHM233 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. LUHM233 Supplier R&D Systems™ Supplier No. LUHM233

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

- Magnetic format allows for easier wash steps - Undergoes similar validation testing as Quantikine™ ELISA Kits - Precise and Specific - Requires a small sample volume (<50μL) - Run your assay in just 3-5 hours - Mass-calibrated standards for consistent results with every new lot of material - Enough reagents for one 96-well plate

Specifications

Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target FGF basic/FGF2
Bead Type Magnetic
Assay Sensitivity 4.91pg/mL
Assay Range 24.00 - 5750pg/mL
Gene ID (Entrez) 2247
Gene Symbol FGF2
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum
Sample Volume Cell Culture Supernates (50μL),Serum (13μL),EDTA Plasma (13μL)
Synonym Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Kit Contents
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.