Learn More
Description
Specifications
Specifications
| Assay Range | 10.00 - 640pg/mL |
| Assay Sensitivity | 3pg/mL |
| Conjugate | HRP |
| Cross Reactivity | Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules |
| Product Line | R&D Systems |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Serum |
| Specificity | Natural and Recombinant Human FGF basic. This assay cross-reacts 40% with natural Bovine FGF-basic, 0.15% with natural Bovine FGF-acidic, 17% with Recombinant Bovine FGF-basic, 18% with Recombinant Mouse FGF-basic and 57% with Recombinant Rat FGF-basic. |
| Sufficient For | 1 x 96-well Plate |
| Target Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
