Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human FGF basic/FGF2/bFGF Quantikine ELISA Kit

Catalog No. SFB50
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
SFB50 6 Plates 6 x 96-well Plates
DFB50 1 Plate 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SFB50 Supplier R&D Systems™ Supplier No. SFB50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human FGF basic/FGF2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human FGF basic/FGF2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human FGF basic/FGF2 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul).

Specifications

Assay Range 10.00 - 640pg/mL
Assay Sensitivity 3pg/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum
Specificity Natural and Recombinant Human FGF basic. This assay cross-reacts 40% with natural Bovine FGF-basic, 0.15% with natural Bovine FGF-acidic, 17% with Recombinant Bovine FGF-basic, 18% with Recombinant Mouse FGF-basic and 57% with Recombinant Rat FGF-basic.
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 2247
Quantity 6 Plates
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.