Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FGF-BP-1 Recombinant Protein, PeproTech®

Catalog No. 10066100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10066100UG 100 μg
100-66-1MG 1 mg
10066250UG 250 μg
100-66-25UG 25 μg
10066500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 10066100UG Supplier Gibco™ Supplier No. 10066100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human FGF-BP-1 expressed in E.coli is a 24.0 kDa protein containing 212 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGFBP1 encodes a secreted fibroblast growth factor carrier protein. The encoded protein plays a critical role in cell proliferation, differentiation and migration by binding to fibroblast growth factors and potentiating their biological effects on target cells. The encoded protein may also play a role in tumor growth as an angiogenic switch molecule, and expression of this gene has been associated with several types of cancer including pancreatic and colorectal adenocarcinoma. A pseudogene of this gene is also located on the short arm of chromosome 4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14512
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 9982
Molecular Weight (g/mol) 23.9 kDa
Name Human FGF-BP-1
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 17 kDa HBGF-binding protein; 17 kDa heparin-binding growth factor-binding protein; FGF-binding protein 1; FGFBP; FGF-BP; Fgfbp1; FGF-BP1; FGFBP-1; fibroblast growth factor binding protein 1; fibroblast growth factor-binding protein 1; growth factor binding protein-1; growth factor-binding protein 1; HBP17; heparin-binding growth factor binding protein
Common Name FGFBP1
Gene Symbol FGFBP1
Biological Activity Determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors. The expected ED50 for this effect is 1.5-3.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence MKKKVKNGLH SKVVSEQKDT LGNTQIKQKS RPGNKGKFVT KDQANCRWAA TEQEEGISLK VECTQLDHEF SCVFAGNPTS CLKLKDERVY WKQVARNLRS QKDICRYSKT AVKTRVCRKD FPESSLKLVS STLFGNTKPR KEKTEMSPRE HIKGKETTPS SLAVTQTMAT KAPECVEDPD MANQRKTALE FCGETWSSLC TFFLSIVQDT SC
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.