Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FGF11 Partial ORF (NP_004103, 15 a.a. - 103 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89944440 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-440 10 ug
89-944-441 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-944-440 Supplier Abnova™ Supplier No. H00002256Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this gene has not yet been determined. The expression pattern of the mouse homolog implies a role in nervous system development. [provided by RefSeq]

Sequence: VREPGGSRPVSAQRRVCPRGTKSLCQKQLLILLSKVRLCGGRPARPDRGPEPQLKGIVTKLFCRQGFYLQANPDGSIQGTPEDTSSFTH

Specifications

Accession Number NP_004103
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2256
Molecular Weight (g/mol) 35.53kDa
Name FGF11 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen VREPGGSRPVSAQRRVCPRGTKSLCQKQLLILLSKVRLCGGRPARPDRGPEPQLKGIVTKLFCRQGFYLQANPDGSIQGTPEDTSSFTH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FHF3/FLJ16061/MGC102953/MGC45269
Common Name FGF11
Gene Symbol FGF11
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.