Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FGF22 (Q9HCT0) Recombinant Protein

Catalog No. 89944473 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-473 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-473 Supplier Abnova™ Supplier No. P4383
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The mouse homolog of this gene was found to be preferentially expressed in the inner root sheath of the hair follicle, which suggested a role in hair development. [provided by RefSeq]

Sequence: MTPSASRGPRSYPHLEGDVRWRRLFSSTHFFLRVDPGGRVQGTRWRHGQDSILEIRSVHVGVVVIKAVSSGFYVAMNRRGRLYGSRLYTVDCRFRERIEENGHNTYASQRWRRRGQPMFLALDRRGGPRPGGRTRRYHLSAHFLPVLVS

Specifications

Accession Number Q9HCT0
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 27006
Molecular Weight (g/mol) 17.3kDa
Name FGF22 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
Quantity 20 μg
Immunogen MTPSASRGPRSYPHLEGDVRWRRLFSSTHFFLRVDPGGRVQGTRWRHGQDSILEIRSVHVGVVVIKAVSSGFYVAMNRRGRLYGSRLYTVDCRFRERIEENGHNTYASQRWRRRGQPMFLALDRRGGPRPGGRTRRYHLSAHFLPVLVS
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Common Name FGF22
Gene Symbol FGF22
Biological Activity Activity was determined by the dose-dependent proliferation of 4MBr-5 cells, is typically 50-300ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.