Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FGF7 (180 a.a.) Full-lenth Recombinant Protein with His tag expressed in Barley grain (Hordeum vulgare)

Catalog No. 89944491 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-491 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-491 Supplier Abnova™ Supplier No. P5880
Add to Cart
Add to Cart

Used for Func, SDS-PAGE, WB-Re

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein is a potent epithelial cell-specific growth factor, whose mitogenic activity is predominantly exhibited in keratinocytes but not in fibroblasts and endothelial cells. Studies of mouse and rat homologs of this gene implicated roles in morphogenesis of epithelium, reepithelialization of wounds, hair development and early lung organogenesis. [provided by RefSeq]

Specifications

For Use With (Application) Functional Assay, SDS-PAGE, Western Blot
Formulation Lyophilized from PBS, pH 7.2
Gene ID (Entrez) 2252
Molecular Weight (g/mol) 14kDa
Name Human FGF7 (180 a.a.) Full-lenth Recombinant Protein with His tag expressed in Barley grain (Hordeum vulgare)
Purification Method Chromatography
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 1 mg
Storage Requirements Store at −20°C.Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.005ng/μg (0.05EU/μg) as measured by turbidimetric kineticassay.
Gene Alias HBGF-7, KGF
Common Name FGF7
Gene Symbol FGF7
Biological Activity Activity has been measured by cell-based proliferation assay using 4MBr5 rhesus monkey epithelial cells. ED50 is typically below 4ng/mL.
Species Plants
Recombinant Recombinant
Protein Tag His Tag
Expression System Hordeum vulgare (barley) expression system
Form Lyophilized
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.