Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FGFR1 (NM_023110, 399 a.a. - 822 a.a.) Partial Recombinant with GST-tag expressed in Sf9 cells

Catalog No. 89944511 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-511 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-511 Supplier Abnova™ Supplier No. P3862

Used for Func, SDS-PAGE

The protein encoded by this gene is a member of the fibroblast growth factor receptor (FGFR) family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein consists of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member binds both acidic and basic fibroblast growth factors and is involved in limb induction. Mutations in this gene have been associated with Pfeiffer syndrome, Jackson-Weiss syndrome, Antley-Bixler syndrome, osteoglophonic dysplasia, and autosomal dominant Kallmann syndrome 2. Chromosomal aberrations involving this gene are associated with stem cell myeloproliferative disorder and stem cell leukemia lymphoma syndrome. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq]

Specifications

Accession Number NM_023110
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 2260
Molecular Weight (g/mol) 73kDa
Name FGFR1 (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BFGFR/CD331/CEK/FGFBR/FLG/FLJ99988/FLT2/HBGFR/KAL2/N-SAM
Common Name FGFR1
Gene Symbol FGFR1
Biological Activity The specific activity was determined to be 177nmol /min/mg as per Activity Assay Protocol.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Insect cell (Sf9) expression system
Form Liquid
Purity or Quality Grade >90% by densitometry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.