Learn More
Haematologic Technologies Inc HUMAN FIBRINOGEN 2MG
Supplier:  Haematologic Technologies Inc HCI0150R
Encompass_Preferred
Coagulation protein, Human Fibrinogen, 2 milligrams, Purified from plasma, Mode of action: Precursor molecule which is cleaved by thrombin to form fibrin clot, RUO, Supplied in 10 mM sodium citrate, 10 mM sodium phosphate, pH 7.3, Store at -80 C.
Catalog No. 50-883-460
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.