Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fibroblast Activation Protein alpha/FAP PE-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. FAB3715P100
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3715P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3715P100 Supplier R&D Systems Supplier No. FAB3715P100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Flow Cytometry∣Immunocytochemistry∣MACS.

Specifications

Antigen Fibroblast Activation Protein alpha/FAP
Applications Flow Cytometry, Immunocytochemistry, Magnetic Cell Separation
Classification Monoclonal
Clone 427819
Conjugate R-PE
Dilution Flow Cytometry 10 μL/106 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q12884
Gene Alias 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin
Gene Symbols FAP
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Fibroblast Activation Protein alpha/FAP, Leu26-Asp760, Accession # Q12884
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2191
Test Specificity Detects human Fibroblast Activation Protein alpha/FAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DPP6 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Product Type Antibody
Form Supernatant
Isotype IgG1
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.