Learn More
Biomatik Corporation Human Fibroblast Growth Factor 6 (FGF6) CLIA Kit
Supplier:  Biomatik Corporation EKU09721
Encompass_Preferred
Human Fibroblast Growth Factor 6 (FGF6) CLIA Kit for detecting FGF6 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.62pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P10767. Gene ID: 2251. Target Synonyms: HBGF6; HST2; HST2 Oncogene; Heparin Secretory-Transforming Protein 2; Heparin-Binding Growth Factor 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8457
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.