Learn More
Specifications
Specifications
| Quantity | 32 Samples |
| Species | Human |
| Binding Target | Fibronectin |
| Assay Sensitivity | 0.0109 ng/mL |
| Assay Range | 0.03 to 321 ng/mL |
| Gene ID (Entrez) | 2335 |
| Product Line | ProteinSimple |
| Sample Type | Cell Culture Supernates, Serum, EDTA (Plasma), Heparin (Plasma), Saliva |
| Synonym | CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
