Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FKBP6 (NP_003593, 1 a.a. - 327 a.a.) Full-length Recombinant Protein with His tag

Catalog No. 89944615 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-615 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-615 Supplier Abnova™ Supplier No. P3508
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The protein may have cis-trans prolyl isomerase activity, and binds to clathrin heavy chain and heat shock protein 72. This gene is found to be deleted in Williams syndrome, and the orthologous gene in mouse is essential for fertility and homologous pairing in male meiosis. [provided by RefSeq]

Sequence: MGSSHHHHHHSSGLVPRGSHMGGSALNQGVLEGDDAPGQSLYERLSQRMLDISGDRGVLKDVIREGAGDLVAPDASVLVKYSGYLEHMDRPFDSNYFRKTPRLMKLGEDITLWGMELGLLSMRRGELARFLFKPNYAYGTLGCPPLIPPNTTVLFEIELLDFLDCAESDKFCALSAEQQDQFPLQKVLKVAATEREFGNYLFRQNRFYDAKVRYKRALLLLRRRSAPPEEQHLVEAAKLPVLLNLSFTYLKLDRPTIALCYGEQALIIDQKNAKALFRCGQACLLLTEYQKARDFLVRAQKEQPFNHDINNELKKLASCYRDYVDKEKEMWHRMFAPCGDGSTAGES

Specifications

Accession Number NP_003593
Concentration 0.5 mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 8468
Molecular Weight (g/mol) 39.3kDa
Name FKBP6 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing Loading 3 ug protein in 15% SDS-PAGE
Quantity 50 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMGGSALNQGVLEGDDAPGQSLYERLSQRMLDISGDRGVLKDVIREGAGDLVAPDASVLVKYSGYLEHMDRPFDSNYFRKTPRLMKLGEDITLWGMELGLLSMRRGELARFLFKPNYAYGTLGCPPLIPPNTTVLFEIELLDFLDCAESDKFCALSAEQQDQFPLQKVLKVAATEREFGNYLFRQNRFYDAKVRYKRALLLLRRRSAPPEEQHLVEAAKLPVLLNLSFTYLKLDRPTIALCYGEQALIIDQKNAKALFRCGQACLLLTEYQKARDFLVRAQKEQPFNHDINNELKKLASCYRDYVDKEKEMWHRMFAPCGDGSTAGES
Storage Requirements Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FKBP36/MGC87179/PPIase
Common Name FKBP6
Gene Symbol FKBP6
Biological Activity Specific activity is > 190nmoles/min/μg, and is defined as the amount of enzyme that cleaves 1μmole of suc-AAFP-pNA per minute at 25°C in Tris-HCl pH 8.0 using chymotrypsin.
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.