Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human FLRG Quantikine ELISA Kit
SDP

Catalog No. DFLRG0 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DFLRG0 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DFLRG0 Supplier R&D Systems™ Supplier No. DFLRG0
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human FLRG Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human FLRG Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Follistatin-related Gene Protein/FLRG/Fstl3 in cell culture supernates (100 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul), urine (50 ul), human milk (50 ul).

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 8.55pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human FLRG
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 10272
Sample Volume Cell Culture Supernates (100uL),Serum (20uL),EDTA Plasma (20uL),Heparin Plasma (20uL),Urine (50uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym FLRGFSRP, Follistatin-like 3, follistatin-like 3 (secreted glycoprotein), Follistatin-like protein 3, Follistatin-related gene protein, follistatin-related protein 3, FSTL3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.