Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Flt-3 Ligand (FLT3L) Recombinant Protein, PeproTech®

Catalog No. 300192UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-19-2UG 2 μg
30019100UG 100 μg
300-19-10UG 10 μg
300-19-1MG 1 mg
30019250UG 250 μg
30019500UG 500 μg
300-19-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 300-19-2UG Supplier Gibco™ Supplier No. 300192UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Flt3-Ligand is a soluble 17.6 kDa protein consisting of 155 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49771
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 2323
Molecular Weight (g/mol) 17.6 kDa
Name Human Flt-3 Ligand (FLT3L)
Quantity 2 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias FL; Flt 3 ligand; Flt3; Flt3 L; Flt3 ligand; Flt3l; Flt3lg; fms related receptor tyrosine kinase 3 ligand; fms related tyrosine kinase 3 ligand; FMS-like tyrosine kinase 3 ligand; fms-related tyrosine kinase 3; fms-related tyrosine kinase 3 ligand; H-FLT-3-L; Ly72L; M-FLT-3-L; NEWGENE_2322792; SL cytokine
Common Name FLT3LG
Gene Symbol FLT3LG
Biological Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of human AML5 cells is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence TQDCSFQHSP ISSDFAVKIR ELSDYLLQDY PVTVASNLQD EELCGGLWRL VLAQRWMERL KTVAGSKMQG LLERVNTEIH FVTKCAFQPP PSCLRFVQTN ISRLLQETSE QLVALKPWIT RQNFSRCLEL QCQPDSSTLP PPWSPRPLEA TAPTA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.