Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FMOD Full-length ORF (AAH35281, 1 a.a. - 376 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89944961 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-961 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-961 Supplier Abnova™ Supplier No. H00002331P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Fibromodulin is a member of a family of small interstitial proteoglycans, containing a central region composed of leucine-rich repeats with 4 keratan sulfate chains flanked by disulfide-bonded terminal domains. It may participate in the assembly of the extracellular matrix as it interacts with type I and type II collagen fibrils and inhibits fibrillogenesis in vitro. It may also regulate TGF-beta activities by sequestering TGF-beta into the extracellular matrix. [provided by RefSeq]

Sequence: MQWISLLLLAGLFSLSQAQYEDDPHWWFHYLRSQQSTYYDPYDPYPYETYEPYPYGVDEGPAYTYGSPSPPDPRDCPQECDCPPNFPTAMYCDNRNLKYLPFVPSRMKYVYFQNNQITSIQEGVFDNATGLLWIALHGNQITSDKVGRKVFSKLRHLERLYLDHNNLTRMPGPLPRSLRELHLDHNQISRVPNNALEGLENLTALYLQHNEIQEVGSSMRGLRSLILLDLSYNHLRKVPDGLPSALEQLYMEHNNVYTVPDSYFRGAPKLLYVRLSHNSLTNNGLASNTFNSSSLLELDLSYNQLQKIPPVNTNLENLYLQGNRINEFSISSFCTVADVVNFSKLQVLRLDGNEIKRSAMPADAPLCLRLASLIEI

Specifications

Accession Number AAH35281
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2331
Molecular Weight (g/mol) 67.10kDa
Name FMOD (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MQWISLLLLAGLFSLSQAQYEDDPHWWFHYLRSQQSTYYDPYDPYPYETYEPYPYGVDEGPAYTYGSPSPPDPRDCPQECDCPPNFPTAMYCDNRNLKYLPFVPSRMKYVYFQNNQITSIQEGVFDNATGLLWIALHGNQITSDKVGRKVFSKLRHLERLYLDHNNLTRMPGPLPRSLRELHLDHNQISRVPNNALEGLENLTALYLQHNEIQEVGSSMRGLRSLILLDLSYNHLRKVPDGLPSALEQLYMEHNNVYTVPDSYFRGAPKLLYVRLSHNSLTNNGLASNTFNSSSLLELDLSYNQLQKIPPVNTNLENLYLQGNRINEFSISSFCTVADVVNFSKLQVLRLDGNEIKRSAMPADAPLCLRLASLIEI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias SLRR2E
Common Name FMOD
Gene Symbol FMOD
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.