Learn More
R&D Systems™ Human Follistatin-like 4/FSTL4 (NP_055897) VersaClone cDNA
Description
- FSTL4 is a presumably secreted member of the follistatin domaincontaining protein family.
- Follistatin itself is a potent inhibitor of the TGF-beta superfamily member, activin, and this activity is reflected in several other proteins of this family that contain varying numbers of the cysteine-rich follistatin domain.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hFSTL4 |
| Accession Number | NP_055897 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2542 bp |
| Gene ID (Entrez) | 23105 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.