Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Follistatin Quantikine ELISA Kit

Catalog No. DFN00
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DFN00 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DFN00 Supplier R&D Systems™ Supplier No. DFN00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Follistatin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Follistatin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Follistatin in cell culture supernates (10 ul), serum (100 ul), edta plasma (100 ul), follicular fluid (10 ul).

Specifications

Assay Range 250.00 - 16000pg/mL
Assay Sensitivity 83pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Follicular Fluid, Plasma (EDTA), Serum
Specificity Natural and Recombinant Human Follistatin
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 10468
Sample Volume Cell Culture Supernates (10uL),Serum (100uL),EDTA Plasma (100uL),Follicular Fluid (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym follistatin, follistatin isoform FST317, FS, FSActivin-binding protein, FST
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.