Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human FOLR1 Quantikine ELISA Kit
SDP

Catalog No. DFLR10 Shop All R&D Systems Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DFLR10 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DFLR10 Supplier R&D Systems™ Supplier No. DFLR10
Add to Cart
Add to Cart
This item is not returnable. View return policy

a 4.5 hour solid-phase ELISA designed to measure human FOLR1 in cell culture supernates, serum, plasma, saliva, urine, and human milk.

The Human FOLR1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human FOLR1 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), saliva (10 ul), urine (10 ul), human milk (10 ul).

Specifications

Assay Range 46.90 - 3000pg/mL
Assay Sensitivity 14.9pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human FOLR1. This assay recognizes Recombinant Human FOLR1 in the presence of folic acid.
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 2348
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Saliva (10uL),Urine (10uL),Human Milk (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, Folbp1, FOLR, FR-alpha, KB cells FBP, MOv18, Ovarian tumor-associated antigen MOv18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.