Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FOXP4 Full-length ORF (NP_001012427.1, 1 a.a. - 678 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945102 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-102 25 μg
89-945-101 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-102 Supplier Abnova™ Supplier No. H00116113P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Many members of the forkhead box gene family, including members of subfamily P, have roles in mammalian oncogenesis. This gene may play a role in the development of tumors of the kidney and larynx. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms. [provided by RefSeq]

Sequence: MMVESASETIRSAPSGQNGVGSLSGQADGSSGGATGTTASGTGREVTTGADSNGEMSPAELLHFQQQQALQVARQFLLQQASGLSSPGNNDSKQSASAVQVPVSVAMMSPQMLTPQQMQQILSPPQLQALLQQQQALMLQQEYYKKQQEQLHLQLLTQQQAGKPQPKEALGNKQLAFQQQLLQMQQLQQQHLLNLQRQGLVSLQPNQASGPLQTLPQAAVCPTDLPQLWKGEGAPGQPAEDSVKQEGLDLTGTAATATSFAAPPKVSPPLSHHTLPNGQPTVLTSRRDSSSHEETPGSHPLYGHGECKWPGCETLCEDLGQFIKHLNTEHALDDRSTAQCRVQMQVVQQLEIQLAKESERLQAMMAHLHMRPSEPKPFSQPLNPVPGSSSFSKVTVSAADSFPDGLVHPPTSAAAPVTPLRPPGLGSASLHGGGPARRRSSDKFCSPISSELAQNHEFYKNADVRPPFTYASLIRQAILETPDRQLTLNEIYNWFTRMFAYFRRNTATWKNAVRHNLSLHKCFVRVENVKGAVWTVDEREYQKRRPPKMTGSPTLVKNMISGLSYGALNASYQAALAESSFPLLNSPGMLNPGSASSLLPLSHDDVGAPVEPLPSNGSSSPPRLSPPQYSHQVQVKEEPAEAEEDRQPGPPLGAPNPSASGPPEDRDLEEELPGEELS

Specifications

Accession Number NP_001012427.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 116113
Molecular Weight (g/mol) 99.6kDa
Name FOXP4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MMVESASETIRSAPSGQNGVGSLSGQADGSSGGATGTTASGTGREVTTGADSNGEMSPAELLHFQQQQALQVARQFLLQQASGLSSPGNNDSKQSASAVQVPVSVAMMSPQMLTPQQMQQILSPPQLQALLQQQQALMLQQEYYKKQQEQLHLQLLTQQQAGKPQPKEALGNKQLAFQQQLLQMQQLQQQHLLNLQRQGLVSLQPNQASGPLQTLPQAAVCPTDLPQLWKGEGAPGQPAEDSVKQEGLDLTGTAATATSFAAPPKVSPPLSHHTLPNGQPTVLTSRRDSSSHEETPGSHPLYGHGECKWPGCETLCEDLGQFIKHLNTEHALDDRSTAQCRVQMQVVQQLEIQLAKESERLQAMMAHLHMRPSEPKPFSQPLNPVPGSSSFSKVTVSAADSFPDGLVHPPTSAAAPVTPLRPPGLGSASLHGGGPARRRSSDKFCSPISSELAQNHEFYKNADVRPPFTYASLIRQAILETPDRQLTLNEIYNWFTRMFAYFRRNTATWKNAVRHNLSLHKCFVRVENVKGAVWTVDEREYQKRRPPKMTGSPTLVKNMISGLSYGALNASYQAALAESSFPLLNSPGMLNPGSASSLLPLSHDDVGAPVEPLPSNGSSSPPRLSPPQYSHQVQVKEEPAEAEEDRQPGPPLGAPNPSASGPPEDRDLEEELPGEELS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ40908/FLJ44184/hFKHLA
Common Name FOXP4
Gene Symbol FOXP4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.