Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FPRL1/FPR2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3479
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3479 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3479 Supplier R&D Systems Supplier No. MAB3479
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

FPRL1/FPR2 Monoclonal specifically detects FPRL1/FPR2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen FPRL1/FPR2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 304405
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P25090
Gene Alias ALXR, FMLP-R-I, FMLP-R-II, FMLPX, formyl peptide receptor 2, Formyl peptide receptor-like 1RFP, FPR2, FPR2A, FPRH1FMLP-related receptor I, FPRL1LXA4 receptor, HM63, HM63FPRH2, Lipoxin A4 receptor, lipoxin A4 receptor (formyl peptide receptor related), LXA4 Receptor, LXA4RFMLP-R-I, N-formyl peptide receptor 2, RFP
Gene Symbols FPR2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human FPRL1 Met1-Met351 Accession # P25090
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2358
Test Specificity Stains FPRL1 transfectants but not irrelevant transfectants. It also stains monocytes and granulocytes.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.