Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Fractalkine (CX3CL1) Recombinant Protein, PeproTech®

Catalog No. 30031250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30031250UG 250 μg
30031100UG 100 μg
300-31-1MG 1 mg
300-31-20UG 20 μg
30031500UG 500 μg
300-31-50UG 50 μg
300-31-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 30031250UG Supplier Gibco™ Supplier No. 30031250UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Fractalkine is an 8.5 kDa protein containing 76 amino acid residues, including the four conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Chemokines are a family of proteins associated with the trafficking of leukocytes in immune surveillance and inflammatory cell recruitment. They are classified based on the positions of key cysteine residues. CX3CL1 is a CX3C chemokine known to induce adhesion and migration of leukocytes mediated by a membrane-bound and soluble form respectively. Recent experiments have shown that CX3CL1 can suppress the production of nitrous oxide, interleukin-6, and TNF-a in activated microglia and neuronal cells, suggesting that it may act as an intrinsic inhibitor against neurotoxicity by activated microglia. Its receptor, CX3CR1, also functions as a co-receptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by CX3CL1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78423
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6376
Molecular Weight (g/mol) 8.5 kDa
Name Human Fractalkine (CX3CL1)
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias A-152E5.2; AB030188; ABCD-3; Acc1; AI848747; C3Xkine; chemokine (C-X3-C motif) ligand 1; CX3C; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1; C-X3-C motif chemokine ligand 1; Cx3cl1; CXC3; CXC3C; D8Bwg0439e; Fkn; Fractalkine; neurotactin; NTN; NTT; Processed fractalkine; SCYD1; small inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, neurotactin); small inducible cytokine subfamily D (Cys-X3-Cys), member-1; small inducible cytokine subfamily D, 1; small-inducible cytokine D1
Common Name CX3CL1
Gene Symbol Cx3cl1
Biological Activity Determined by its ability to chemoattract human T cells using a concentration range of 5.0-10.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence QHHGVTKCNI TCSKMTSKIP VALLIHYQQN QASCGKRAII LETRQHRLFC ADPKEQWVKD AMQHLDRQAA ALTRNG
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.