Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FRP-1 (soluble) Recombinant Protein, PeproTech®

Catalog No. 1202920UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-29-20UG 20 μg
12029100UG 100 μg
120-29-1MG 1 mg
12029250UG 250 μg
12029500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 120-29-20UG Supplier Gibco™ Supplier No. 1202920UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human sFRP-1 is a glycosylated 283 amino acid protein containing a cysteine-rich Frizzled homologous domain. The calculated molecular weight of Recombinant Human sFRP-1 is 32.5 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Secreted frizzled-related protein 1 (SFRP1) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP1 and SFRP5 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP1 is expressed in several human tissues, with the highest levels in heart.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N474
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 6422
Molecular Weight (g/mol) 32.5 kDa
Name Human FRP-1 (soluble)
Quantity 20 μg
Source HeLa cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 2210415K03Rik; AW011917; AW107218; AW742929; frizzled-related protein; FRP; FRP1; FRP-1; FrzA; SARP2; SARP-2; Secreted apoptosis-related protein 2; secreted frizzled related protein 1; secreted frizzled-related protein 1; secreted frizzled-related sequence protein 1; SFRP1; sFRP-1
Common Name SFRP1
Gene Symbol SFRP1
Biological Activity Determined by its ability to inhibit BMP-2 proliferation effect of ATDC-5 cells. The expected ED50 is 0.3-0.5 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence SEYDYVSFQS DIGPYQSGRF YTKPPQCVDI PADLRLCHNV GYKKMVLPNL LEHETMAEVK QQASSWVPLL NKNCHAGTQV FLCSLFAPVC LDRPIYPCRW LCEAVRDSCE PVMQFFGFYW PEMLKCDKFP EGDVCIAMTP PNATEASKPQ GTTVCPPCDN ELKSEAIIEH LCASEFALRM KIKEVKKENG DKKIVPKKKK PLKLGPIKKK DLKKLVLYLK NGADCPCHQL DNLSHHFLIM GRKVKSQYLL TAIHKWDKKN KEFKNFMKKM KNHECPTFQS VFK
Content And Storage -20°C
Expression System HeLa cells
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.