Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FRP-4 (soluble) Recombinant Protein, PeproTech®

Catalog No. 12050500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12050500UG 500 μg
12050100UG 100 μg
120-50-1MG 1 mg
12050250UG 250 μg
120-50-25UG 25 μg
120-50-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 12050500UG Supplier Gibco™ Supplier No. 12050500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sFRP-4 is a glycoprotein of 328 amino acid residues that contains a cysteine-rich, Frizzled-homologous domain. The calculated molecular weight of CHO cell-derived Recombinant Human sFRP-4 is 37.8 kDa; however, due to glycosylation, the protein migrates at an apparent molecular weight of approximately 55 - 65 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Secreted Frizzled-Related Proteins (sFRPs) are a family of glycosylated Wnt antagonists characterized by a conserved cysteine-rich domain that shares homology with the cysteine-rich, extracellular domain Frizzled proteins use for the binding of Wnt proteins and receptors. Lacking the transmembrane and intracellular domains of the Frizzled proteins, sFRPs function as soluble modulators of the Wnt signaling pathway through the direct binding of Wnt proteins to this cysteine-rich domain, and the resultant inhibition of Wnt receptor binding and signaling capabilities. sFRP-4 is widely distributed in a variety of embryonic and adult tissues where it can function as a circulating antiangiogenic factor, a potent proapoptotic factor, an inhibitor of insulin secretion, and a suppressor of both tumor growth and metastatic potential through disruption of the Wnt signaling pathway. Research has demonstrated the existence of a direct correlation between the downregulation and/or absence of circulating sFRP-4 and the progression of several cancer types, including ovarian, endometrial, prostate and lung. Upregulation of circulating sFRP-4 has been linked to the deterioration of glucose metabolism in the case of type 2 diabetes, as well as the suppression of the keratinocyte hyperproliferation and epidermal hyperlasia that are definitive of psoriasis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6FHJ7
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 6424
Molecular Weight (g/mol) 37.8 kDa
Name Human FRP-4 (soluble)
Quantity 500 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Ddc4; DDC-4 protein; Frizzled protein, human endometrium; frizzled related protein; frizzled-related protein; frizzled-related protein 4; FRP; FRP-4; FRPHE; MGC26498; secreted frizzled related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related sequence protein 4; Sfrp4; sFRP-4
Common Name SFRP4
Gene Symbol SFRP4
Biological Activity Determined by its ability to decrease alkaline phosphatase activity in CCL-226 cells when treated with 25 ng/ml of Murine Wnt-3 a.
Conjugate Unconjugated
Recombinant Recombinant
Sequence VRGAPCEAVR IPMCRHMPWN ITRMPNHLHH STQENAILAI EQYEELVDVN CSAVLRFFLC AMYAPICTLE FLHDPIKPCK SVCQRARDDC EPLMKMYNHS WPESLACDEL PVYDRGVCIS PEAIVTDLPE DVKWIDITPD MMVQERPLDV DCKRLSPDRC KCKKVKPTLA TYLSKNYSYV IHAKIKAVQR SGCNEVTTVV DVKEIFKSSS PIPRTQVPLI TNSSCQCPHI LPHQDVLIMC YEWRSRMMLL ENCLVEKWRD QLSKRSIQWE ERLQEQRRTV QDKKKTAGRT SRSNPPKPKG KPPAPKPASP KKNIKTRSAQ KRTNPKRV
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.