Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FRP-5 (soluble) Recombinant Protein, PeproTech®

Catalog No. 1205310UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-53-10UG 10 μg
12053100UG 100 μg
120-53-1MG 1 mg
12053250UG 250 μg
12053500UG 500 μg
120-53-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 120-53-10UG Supplier Gibco™ Supplier No. 1205310UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sFRP5 is a 288-amino-acid length glycoprotein with a calculated molecular weight of 32.7 kDa; however, due to glycosylation, protein migration occurs at an apparent molecular weight of approximately 33-35 kDa by SDS-PAGE analysis under both reducing and non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Secreted frizzled-related protein 5 (SFRP5) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP5 and SFRP1 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP5 is highly expressed in the retinal pigment epithelium, and moderately expressed in the pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5T4F7
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 6425
Molecular Weight (g/mol) 32.7 kDa
Name Human FRP-5 (soluble)
Quantity 10 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI605071; Frizzled-related protein 1 b; FRP1B; FRP-1 b; SARP3; SARP-3; secreted apoptosis related protein 3; secreted apoptosis-related protein 3; secreted frizzled related protein 5; secreted frizzled-related protein 5; secreted frizzled-related sequence protein 5; SFRP5; sFRP-5
Common Name SFRP5
Gene Symbol SFRP5
Biological Activity Determined by its ability to decrease alkaline phosphatase activity in CCL-226 cells when treated with 25 ng/ml of Murine Wnt-3 a.
Conjugate Unconjugated
Recombinant Recombinant
Sequence EEYDYYGWQA EPLHGRSYSK PPQCLDIPAD LPLCHTVGYK RMRLPNLLEH ESLAEVKQQA SSWLPLLAKR CHSDTQVFLC SLFAPVCLDR PIYPCRSLCE AVRAGCAPLM EAYGFPWPEM LHCHKFPLDN DLCIAVQFGH LPATAPPVTK ICAQCEMEHS ADGLMEQMCS SDFVVKMRIK EIKIENGDRK LIGAQKKKKL LKPGPLKRKD TKRLVLHMKN GAGCPCPQLD SLAGSFLVMG RKVDGQLLLM AVYRWDKKNK EMKFAVKFMF SYPCSLYYPF FYGAAEPH
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.