Learn More
ILEX LIFE SCIENCES LLC HUMAN FSH HEK293 RECOMB 1 MG
SDP
Supplier:  ILEX LIFE SCIENCES LLC A22531MG
Encompass_Preferred
Ilex Life Sciences Human Follicle Stimulating Hormone (FSH), HEK293 Recombinant Protein, 1 mg. DESCRIPTION: FSH Human, HEK293 Recombinant is a heterodimeric, glycosylated, polypeptide chain transfected with two expression plasmids encoding the human FSH-alpha chain (#P01215) (Ala25-Ser116) and human FSH-beta chain (#P01225) (Asn19-Glu129); total Mw of 38 kDa. BIOACTIVITY: The ED50 as determined by cAMP accumulation in human FSH Receptor transfected CHO cells was found to be 80-450 pg/ml. FORMULATION: Lyophilized from 0.2µm sterile filtered solution containing PBS, pH 7.4. PURITY: Greater than 95.0% as determined by SDS-PAGE. USAGE: This product is intended for Laboratory Research Use Only. Not for use in diagnostic or therapeutic procedures.
Catalog No. 50-273-4011
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.