Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FZD4 Full-length ORF (NP_036325.2, 1 a.a. - 537 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945291 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-291 25 μg
89-945-290 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-291 Supplier Abnova™ Supplier No. H00008322P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. This protein may play a role as a positive regulator of the Wingless type MMTV integration site signaling pathway. A transcript variant retaining intronic sequence and encoding a shorter isoform has been described, however, its expression is not supported by other experimental evidence. [provided by RefSeq]

Sequence: MAWRGAGPSVPGAPGGVGLSLGLLLQLLLLLGPARGFGDEEERRCDPIRISMCQNLGYNVTKMPNLVGHELQTDAELQLTTFTPLIQYGCSSQLQFFLCSVYVPMCTEKINIPIGPCGGMCLSVKRRCEPVLKEFGFAWPESLNCSKFPPQNDHNHMCMEGPGDEEVPLPHKTPIQPGEECHSVGTNSDQYIWVKRSLNCVLKCGYDAGLYSRSAKEFTDIWMAVWASLCFISTAFTVLTFLIDSSRFSYPERPIIFLSMCYNIYSIAYIVRLTVGRERISCDFEEAAEPVLIQEGLKNTGCAIIFLLMYFFGMASSIWWVILTLTWFLAAGLKWGHEAIEMHSSYFHIAAWAIPAVKTIVILIMRLVDADELTGLCYVGNQNLDALTGFVVAPLFTYLVIGTLFIAAGLVALFKIRSNLQKDGTKTDKLERLMVKIGVFSVLYTVPATCVIACYFYEISNWALFRYSADDSNMAVEMLKIFMSLLVGITSGMWIWSAKTLHTWQKCSNRLVNSGKVKREKRGNGWVKPGKGSETVV

Specifications

Accession Number NP_036325.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8322
Molecular Weight (g/mol) 86.3kDa
Name FZD4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MAWRGAGPSVPGAPGGVGLSLGLLLQLLLLLGPARGFGDEEERRCDPIRISMCQNLGYNVTKMPNLVGHELQTDAELQLTTFTPLIQYGCSSQLQFFLCSVYVPMCTEKINIPIGPCGGMCLSVKRRCEPVLKEFGFAWPESLNCSKFPPQNDHNHMCMEGPGDEEVPLPHKTPIQPGEECHSVGTNSDQYIWVKRSLNCVLKCGYDAGLYSRSAKEFTDIWMAVWASLCFISTAFTVLTFLIDSSRFSYPERPIIFLSMCYNIYSIAYIVRLTVGRERISCDFEEAAEPVLIQEGLKNTGCAIIFLLMYFFGMASSIWWVILTLTWFLAAGLKWGHEAIEMHSSYFHIAAWAIPAVKTIVILIMRLVDADELTGLCYVGNQNLDALTGFVVAPLFTYLVIGTLFIAAGLVALFKIRSNLQKDGTKTDKLERLMVKIGVFSVLYTVPATCVIACYFYEISNWALFRYSADDSNMAVEMLKIFMSLLVGITSGMWIWSAKTLHTWQKCSNRLVNSGKVKREKRGNGWVKPGKGSETVV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CD344/EVR1/FEVR/FZD4S/Fz-4/FzE4/GPCR/MGC34390
Common Name FZD4
Gene Symbol FZD4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.