Learn More
Specifications
Specifications
| Assay Sensitivity | 3.57 pg/mL |
| Product Line | R&D Systems |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Heparin Plasma, Serum |
| Specificity | Natural and Recombinant Human G-CSF |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1440 |
| Measuring Range | 39.10 to 2500 pg/mL |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
