Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human gAcrp30/Adipolean Recombinant Protein, PeproTech®

Catalog No. 4502101M
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-021-01M 2 x 500 μg
45021250UG 250 μg
450-21-25UG 25 μg
45021500UG 500 μg
450-21-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 45-021-01M Supplier Gibco™ Supplier No. 4502101M
Add to Cart
Add to Cart

Recombinant Protein

450-21-1MG will be provided as 2 x 500 μg (450-21-500UG). Recombinant Human gAcrp30/Adipolean is a 16.6 kDa protein consisting of 145 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

gAcrp30 is a naturally occurring globular protein obtained by proteolytic processing of adiponectin. Adiponectin is produced and secreted exclusively by adipocytes, and is a relatively abundant plasma protein, accounting for up to 0.05% of total serum protein. Like Adiponectin, gAcrp30 is capable of decreasing hyperglycemia and reversing insulin resistance. Additionally, gAcrp30 has been shown to be an important factor in promoting fat loss by signaling muscle to absorb and burn Free-Fatty Acids (FFAs). The signaling receptors for adiponectin and gAcrp30 have recently been identified and named AdipoR1 and AdipoR2. AdipoR2 is predominantly expressed in the liver.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15848
For Use With (Application) Control, Western Blot Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 9370
Molecular Weight (g/mol) 16.6 kDa
Name Human gAcrp30/Adipolean
Purification Method Purified
Quantity 2 x 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <2 EU/μg
Gene Alias gAcrp30
Common Name Adipolean
Gene Symbol ADIPOQ
Biological Activity Determined by its ability to inhibit the proliferation of murine M1 cells. The expected ED50 for this effect is 1.0-3.0 ug/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN
Content And Storage -20°C
Activity Determined by its ability to inhibit the proliferation of murine M1 cells. The expected ED50 for this effect is 1.0-3.0 ug/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.