Learn More
Human Galanin R1/GALR1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Galanin R1/GALR1 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 613207 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. |
| Gene Accession No. | P47211 |
| Gene Alias | GAL1, galanin receptor 1, galanin receptor type 1, GalaninR1, GALNR1, GALNRGALNR1GAL1-R, GALR1, GALR-1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.