Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GALC Full-length ORF (AAH36518, 27 a.a. - 669 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945426 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-426 10 ug
89-945-427 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-426 Supplier Abnova™ Supplier No. H00002581P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a lysosomal protein which hydrolyzes the galactose ester bonds of galactosylceramide, galactosylsphingosine, lactosylceramide, and monogalactosyldiglyceride. Mutations in this gene have been associated with Krabbe disease, also known as globoid cell leukodystrophy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: YVLDDSDGLGREFDGIGAVSGGGATSRLLVNYPEPYRSQILDYLFKPNFGASLHILKVEIGGDGQTTDGTEPSHMHYALDGNYFRGYEWWLMKEAKKRNPNITLIGLPWSFPGWLGKGFDWPYVNLQLTAYYVVTWIVGAKRYHDLDIDYIGIWNERSYNANYIKILRKMLNYQGLQRVKIIASDNLWESISASMLLDAELFKVVDVIGAHYPGTHSAKDAKLTGKKLWSSEDFSTLNSDMGAGCWGRILNQNYINGYMTSTIAWNLVASYYEQLPYGRCGLMTAQEPWSGHYVVESPVWVSAHTTQFTQPGWYYLKTVGHLEKGGSYVALTDGLGNLTIIIETMSHKHSKCIRPFLPYFNVSQQFATFVLKGSFSEIPELQVWYTKLGKTSERFLFKQLDSLWLLDSDGSFTLSLHEDELFTLTTLTTGRKGSYPLPPKSQPFPSTYKDDFNVDYPFFSEAPNFADQTGVFEYFTNIEDPGEHHFTLRQVLNQRPITWAADASNTISIIGDYNWTNLTTKCDVYIETPDTGGVFIAGRVNKGGILIRSARGIFFWIFANGSYRVTGDLAGWIIYALGRVEVTAKKWYTLTLTIKGHFASGMLNDKSLWTDIPVNFPKNGWAAIGTHSFEFAQFDNFRVEATR

Specifications

Accession Number AAH36518
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2581
Molecular Weight (g/mol) 96.47kDa
Name GALC (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen YVLDDSDGLGREFDGIGAVSGGGATSRLLVNYPEPYRSQILDYLFKPNFGASLHILKVEIGGDGQTTDGTEPSHMHYALDGNYFRGYEWWLMKEAKKRNPNITLIGLPWSFPGWLGKGFDWPYVNLQLTAYYVVTWIVGAKRYHDLDIDYIGIWNERSYNANYIKILRKMLNYQGLQRVKIIASDNLWESISASMLLDAELFKVVDVIGAHYPGTHSAKDAKLTGKKLWSSEDFSTLNSDMGAGCWGRILNQNYINGYMTSTIAWNLVASYYEQLPYGRCGLMTAQEPWSGHYVVESPVWVSAHTTQFTQPGWYYLKTVGHLEKGGSYVALTDGLGNLTIIIETMSHKHSKCIRPFLPYFNVSQQFATFVLKGSFSEIPELQVWYTKLGKTSERFLFKQLDSLWLLDSDGSFTLSLHEDELFTLTTLTTGRKGSYPLPPKSQPFPSTYKDDFNVDYPFFSEAPNFADQTGVFEYFTNIEDPGEHHFTLRQVLNQRPITWAADASNTISIIGDYNWTNLTTKCDVYIETPDTGGVFIAGRVNKGGILIRSARGIFFWIFANGSYRVTGDLAGWIIYALGRVEVTAKKWYTLTLTIKGHFASGMLNDKSLWTDIPVNFPKNGWAAIGTHSFEFAQFDNFRVEATR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name GALC
Gene Symbol GALC
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.