Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Galectin-1 Recombinant Protein, PeproTech®

Catalog No. 450391MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
250 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-39-1MG 1 mg
45039100UG 100 μg
45039250UG 250 μg
45039500UG 500 μg
450-39-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 450-39-1MG Supplier Gibco™ Supplier No. 450391MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Galectin-1 is a 14.5 kDa protein containing 134 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09382
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 3956
Molecular Weight (g/mol) 14.5 kDa
Name Human Galectin-1
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 14 kDa laminin-binding protein; 14 kDa lectin; AA410090; beta-galactoside binding protein; beta-galactoside-binding lectin; beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14 kDa; DKFZp686E23103; GAL1; GAL-1; Galaptin; Galbp; galectin 1; galectin 1; LGALS1; Galectin1; galectin-1; galectin-1-like protein; GBP; HBL; H-Galectin-1; HLBP14; HPL; L14; L-14.5; Lactose-binding lectin 1; Lect14; Lectin galactoside-binding soluble 1; lectin, galactose binding, soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; lectin, galactoside-binding, soluble, 1 (galectin 1); Lgals1; LGAS1; OPG-1; pm12; Putative MAPK-activating protein PM12; RL 14.5; similar to galactose-binding lectin; S-Lac lectin 1; unnamed protein product
Common Name Galectin 1
Gene Symbol LGALS1
Biological Activity Determined by its ability to chemoattract human blood monocytes. Chemotactic activity was observed at a concentration of 2.5 ug/ml with a peak response obtained (at 250 ug/ml)
Conjugate Unconjugated
Recombinant Recombinant
Sequence ACGLVASNLN LKPGECLRVR GEVAPDAKSF VLNLGKDSNN LCLHFNPRFN AHGDANTIVC NSKDGGAWGT EQREAVFPFQ PGSVAEVCIT FDQANLTVKL PDGYEFKFPN RLNLEAINYM AADGDFKIKC VAFD
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.