Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Galectin-3 Quantikine ELISA Kit
SDP

Catalog No. SGAL30 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SGAL30 1 Pk. (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SGAL30 Supplier R&D Systems™ Supplier No. SGAL30
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Galectin-3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Galectin-3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Galectin-3 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

Specifications

Assay Range 0.31 to 10 ng/mL
Assay Sensitivity 0.085 ng/mL
Conjugate HRP
Cross Reactivity < 0.5% cross-reactivity observed with available related molecules, Cross-species reactivity observed with 1 or more species tested
Immunoassay Kit Format 96-well Strip Plate
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA, Heparin), Plasma (Heparin), Serum
Specificity Natural and Recombinant Human Galectin-3. This assay cross-reacts 58% with Mouse Galectin-3.
For Use With (Application) ELISA
Gene ID (Entrez) 3958
Gene Symbol LGALS3
Precision
  • Intra-Assay Precision (Precision within an assay) Three samples of known concentration were tested on one plate to assess intra-assay precision
  • Inter-Assay Precision (Precision between assays) Three samples of known concentration were tested in separate assays to assess inter-assay precision
Quantity 1 Pk. (6 Plates)
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3
Target Galectin-3
Total Assay Time 4.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.