Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Galectin-3BP/MAC-2BP Quantikine ELISA Kit
SDP

Catalog No. DGBP30B Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DGBP30B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DGBP30B Supplier R&D Systems™ Supplier No. DGBP30B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Galectin-3BP/MAC-2BP Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Galectin-3BP/MAC-2BP Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Galectin-3BP/MAC-2BP/LGALS3BP.

Specifications

Assay Range 0.39 to 25 ng/mL
Assay Sensitivity 0.161 ng/mL
Conjugate HRP
Cross Reactivity <0.5% cross-reactivity observed with available related molecules. <50% cross-species reactivity observed with species tested.
Immunoassay Kit Format 96-well Strip Plate
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human Galectin-3 BP/MAC-2 BP
For Use With (Application) ELISA
Gene ID (Entrez) 3959 (Human)
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Saliva (10uL),Urine (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym 90 K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, Galectin3BP, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, LGALS3BP, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2 BP, MAC-2-BP, serum protein 90 K, Tumor-associated antigen 90 K
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.