Learn More
ACROBiosystems Human Galectin-9 200ug
Supplier:  ACROBiosystems LG9H82H3200UG
Encompass_Preferred
Biotinylated Human Galectin-9 HisAvitag (LG9-H82H3) is expressed from human 293 cells (HEK293). It contains AA Ala 2 - Thr 323 (Accession # O00182-2).
Catalog No. 50-253-4514
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.