Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GALNT13 Full-length ORF (NP_443149.1, 1 a.a. - 556 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945461 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-461 10 μg
89-945-462 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-461 Supplier Abnova™ Supplier No. H00114805P01S

Used for AP, Array, ELISA, WB-Re

The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins (see MUC3A, MIM 158371) by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.[supplied by OMIM]

Sequence: MRRSVYCKVVLATSLMWVLVDVFLLLYFSECNKCDDKKERSLLPALRAVISRNQEGPGEMGKAVLIPKDDQEKMKELFKINQFNLMASDLIALNRSLPDVRLEGCKTKVYPDELPNTSVVIVFHNEAWSTLLRTVYSVINRSPHYLLSEVILVDDASERDFLKLTLENYVKNLEVPVKIIRMEERSGLIRARLRGAAASKGQVITFLDAHCECTLGWLEPLLARIKEDRKTVVCPIIDVISDDTFEYMAGSDMTYGGFNWKLNFRWYPVPQREMDRRKGDRTLPVRTPTMAGGLFSIDRNYFEEIGTYDAGMDIWGGENLEMSFRIWQCGGSLEIVTCSHVGHVFRKATPYTFPGGTGHVINKNNRRLAEVWMDEFKDFFYIISPGVVKVDYGDVSVRKTLRENLKCKPFSWYLENIYPDSQIPRRYYSLGEIRNVETNQCLDNMGRKENEKVGIFNCHGMGGNQVFSYTADKEIRTDDLCLDVSRLNGPVIMLKCHHMRGNQLWEYDAERLTLRHVNSNQCLDEPSEEDKMVPTMQDCSGSRSQQWLLRNMTLGT

Specifications

Accession Number NP_443149.1
For Use With (Application) Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 114805
Molecular Weight (g/mol) 90.4kDa
Name Human GALNT13 Full-length ORF (NP_443149.1, 1 a.a. - 556 a.a.) Recombinant Protein with GST-tag at N-terminal
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ16031, FLJ41157, GalNAc-T13, H_NH0187G20.1, KIAA1918, MGC119459, MGC119461, WUGSC:H_NH0187G20.1
Common Name GALNT13
Gene Symbol GALNT13
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST Tag
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.