Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GALNT7 Full-length ORF (AAH46129.1, 1 a.a. - 657 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945487 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-487 25 μg
89-945-486 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-487 Supplier Abnova™ Supplier No. H00051809P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes GalNAc transferase 7, a member of the GalNAc-transferase family. The enzyme encoded by this gene controls the initiation step of mucin-type O-linked protein glycosylation and transfer of N-acetylgalactosamine to serine and threonine amino acid residues. This enzyme is a type II transmembrane protein and shares common sequence motifs with other family members. Unlike other family members, this enzyme shows exclusive specificity for partially GalNAc-glycosylated acceptor substrates and shows no activity with non-glycosylated peptides. This protein may function as a follow-up enzyme in the initiation step of O-glycosylation. [provided by RefSeq]

Sequence: MRLKIGFILRSLLVVGSFLGLVVLWSSLTPRPDDPSPLSRMREDRDVNDPMPNRGGNGLAPGEDRFKPVVPWPHVEGVEVDLESIRRINKAKNEQEHHAGGDSQKDIMQRQYLTFKPQTFTYHDPVLRPGILGNFEPKEPEPPGVVGGPGEKAKPLVLGPEFKQAIQASIKEFGFNMVASDMISLDRSVNDLRQEECKYWHYDENLLTSSVVIVFHNEGWSTLMRTVHSVIKRTPRKYLAEIVLIDDFSNKEHLKEKLDEYIKLWNGLVKVFRNERREGLIQARSIGAQKAKLGQVLIYLDAHCEVAVNWYAPLVAPISKDRTICTVPLIDVINGNTYEIIPQGGGDEDGYARGAWDWSMLWKRVPLTPQEKRLRKTKTEPYRSPAMAGGLFAIEREFFFELGLYDPGLQIWGGENFEISYKIWQCGGKLLFVPCSRVGHIYRLEGWQGNPPPIYVGSSPTLKNYVRVVEVWWDEYKDYFYASRPESQALPYGDISELKKFREDHNCKSFKWFMEEIAYDITSHYPLPPKNVDWGEIRGFETAYCIDSMGKTNGGFVELGPCHRMGGNQLFRINEANQLMQYDQCLTKGADGSKVMITHCNLNEFKEWQYFKNLHRFTHIPSGKCLDRSEVLHQVFISNCDSSKTTQKWEMNNIHSV

Specifications

Accession Number AAH46129.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51809
Molecular Weight (g/mol) 101.8kDa
Name GALNT7 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MRLKIGFILRSLLVVGSFLGLVVLWSSLTPRPDDPSPLSRMREDRDVNDPMPNRGGNGLAPGEDRFKPVVPWPHVEGVEVDLESIRRINKAKNEQEHHAGGDSQKDIMQRQYLTFKPQTFTYHDPVLRPGILGNFEPKEPEPPGVVGGPGEKAKPLVLGPEFKQAIQASIKEFGFNMVASDMISLDRSVNDLRQEECKYWHYDENLLTSSVVIVFHNEGWSTLMRTVHSVIKRTPRKYLAEIVLIDDFSNKEHLKEKLDEYIKLWNGLVKVFRNERREGLIQARSIGAQKAKLGQVLIYLDAHCEVAVNWYAPLVAPISKDRTICTVPLIDVINGNTYEIIPQGGGDEDGYARGAWDWSMLWKRVPLTPQEKRLRKTKTEPYRSPAMAGGLFAIEREFFFELGLYDPGLQIWGGENFEISYKIWQCGGKLLFVPCSRVGHIYRLEGWQGNPPPIYVGSSPTLKNYVRVVEVWWDEYKDYFYASRPESQALPYGDISELKKFREDHNCKSFKWFMEEIAYDITSHYPLPPKNVDWGEIRGFETAYCIDSMGKTNGGFVELGPCHRMGGNQLFRINEANQLMQYDQCLTKGADGSKVMITHCNLNEFKEWQYFKNLHRFTHIPSGKCLDRSEVLHQVFISNCDSSKTTQKWEMNNIHSV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GALNAC-T7/GalNAcT7
Common Name GALNT7
Gene Symbol GALNT7
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.