Learn More
Abnova™ Human GALNT9 Full-length ORF (NP_068580.2, 1 a.a. - 237 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. This gene is expressed specifically in the brain, with highest expression in the cerebellum. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
| Accession Number | NP_068580.2 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 50614 |
| Molecular Weight (g/mol) | 53.6kDa |
| Name | GALNT9 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | MEVLPCSRVAHIERTRKPYNNDIDYYAKRNALRAAEVWMDDFKSHVYMAWNIPMSNPGVDFGDVSERLALRQRLKCRSFKWYLENVYPEMRVYNNTLTYGEVRNSKASAYCLDQGAEDGDRAILYPCHGMSSQLVRYSADGLLQLGPLGSTAFLPDSKCLVDDGTGRMPTLKKCEDVARPTQRLWDFTQSGPIVSRATGRCLEVEMSKDANFGLRLVVQRCSGQKWMIRNWIKHARH |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.