Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human gamma-Synuclein Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067940
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5745RSP 25 μg
MAB5745R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5745RSP Supplier R&D Systems Supplier No. MAB5745RSP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

gamma-Synuclein Monoclonal specifically detects gamma-Synuclein in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen gamma-Synuclein
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 514304R
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 1-25 ug/mL
Formulation Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6FHG5
Gene Alias BCSG1, BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, PRSN, SNCG, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1), Synuclein-gamma
Gene Symbols SNCG
Host Species Mouse
Immunogen E. coli-derived recombinant human gamma-Synuclein Asn47-Glu120 Accession # Q6FHG5
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6623
Test Specificity Detects human gamma-Synuclein in direct ELISAs and Western blots.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.