Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GASP-1 ELISA Kit

Catalog No. EH209RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH209RB 96 Tests
EH209RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH209RB Supplier Invitrogen™ Supplier No. EH209RB
Add to Cart
Add to Cart

ELISA

Human GASP-1 quantitates human GASP-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GASP-1.

Modulates lysosomal sorting and functional down-regulation of a variety of G-protein coupled receptors. Targets receptors for degradation in lysosomes via its interaction with BECN2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2210415K24Rik; 3110031O14Rik; C87852; G protein-coupled receptor associated sorting protein 1; GASP; GASP1; GASP-1; GPRASP1; G-protein coupled receptor-associated sorting protein 1; Kiaa0443; Per1 interacting protein; per1-interacting protein; Pips
Assay Range 20 to 5,000 pg/mL
Assay Sensitivity 20 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 9737
Gene Symbol GPRASP1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target GPRASP1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.