Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GATA-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7161259
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB26062SP 25 μg
MAB26062100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB26062SP Supplier R&D Systems Supplier No. MAB26062SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GATA-4 Monoclonal specifically detects GATA-4 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen GATA-4
Applications Immunocytochemistry
Classification Monoclonal
Clone 532025
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 μ/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P43694
Gene Alias GATA binding protein 4, GATA4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4
Host Species Mouse
Immunogen E. coli-derived recombinant human GATA-4, Met27-Ser212, Accession No. P43694
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 2626
Test Specificity Detects human GATA-4 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.