Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GCDH Full-length ORF (NP_000150.1, 1 a.a. - 438 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945642 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-642 25 ug
89-945-641 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-642 Supplier Abnova™ Supplier No. H00002639P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family. It catalyzes the oxidative decarboxylation of glutaryl-CoA to crotonyl-CoA and CO(2) in the degradative pathway of L-lysine, L-hydroxylysine, and L-tryptophan metabolism. It uses electron transfer flavoprotein as its electron acceptor. The enzyme exists in the mitochondrial matrix as a homotetramer of 45-kD subunits. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MALRGVSVRLLSRGPGLHVLRTWVSSAAQTEKGGRTQSQLAKSSRPEFDWQDPLVLEEQLTTDEILIRDTFRTYCQERLMPRILLANRNEVFHREIISEMGELGVLGPTIKGYGCAGVSSVAYGLLARELERVDSGYRSAMSVQSSLVMHPIYAYGSEEQRQKYLPQLAKGELLGCFGLTEPNSGSDPSSMETRAHYNSSNKSYTLNGTKTWITNSPMADLFVVWARCEDGCIRGFLLEKGMRGLSAPRIQGKFSLRASATGMIIMDGVEVPEENVLPGASSLGGPFGCLNNARYGIAWGVLGASEFCLHTARQYALDRMQFGVPLARNQLIQKKLADMLTEITLGLHACLQLGRLKDQDKAAPEMVSLLKRNNCGKALDIARQARDMLGGNGISDEYHVIRHAMNLEAVNTYEGTHDIHALILGRAITGIQAFTASK

Specifications

Accession Number NP_000150.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2639
Molecular Weight (g/mol) 74.5kDa
Name GCDH (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MALRGVSVRLLSRGPGLHVLRTWVSSAAQTEKGGRTQSQLAKSSRPEFDWQDPLVLEEQLTTDEILIRDTFRTYCQERLMPRILLANRNEVFHREIISEMGELGVLGPTIKGYGCAGVSSVAYGLLARELERVDSGYRSAMSVQSSLVMHPIYAYGSEEQRQKYLPQLAKGELLGCFGLTEPNSGSDPSSMETRAHYNSSNKSYTLNGTKTWITNSPMADLFVVWARCEDGCIRGFLLEKGMRGLSAPRIQGKFSLRASATGMIIMDGVEVPEENVLPGASSLGGPFGCLNNARYGIAWGVLGASEFCLHTARQYALDRMQFGVPLARNQLIQKKLADMLTEITLGLHACLQLGRLKDQDKAAPEMVSLLKRNNCGKALDIARQARDMLGGNGISDEYHVIRHAMNLEAVNTYEGTHDIHALILGRAITGIQAFTASK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ACAD5/GCD
Common Name GCDH
Gene Symbol GCDH
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.