Learn More
ACROBiosystems Human GCGR 20ug
Supplier:  ACROBiosystems GCRH52P520UG
Encompass_Preferred
Human GCGR Full Length Protein (VLP) (GCR-H52P5) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Asn 432 (Accession # P47871-1).
Catalog No. 50-253-5034
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.