Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human GDF-15 Quantikine ELISA Kit
GSA_VASDP

Catalog No. SGD150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SGD150 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SGD150 Supplier R&D Systems™ Supplier No. SGD150

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human GDF-15 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human GDF-15 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human GDF-15 in cell culture supernates (50 ul), serum (13 ul), edta plasma (13 ul), heparin plasma (13 ul), urine (10 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 23.40 - 1500pg/mL
Assay Sensitivity 4.39pg/mL
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 0.5% cross-reactivity observed with available related molecules, Cross-species reactivity not tested
Immunoassay Kit Format 96-well Strip Plate
Product Type Human GDF-15 Quantikine ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human GDF-15
For Use With (Application) ELISA
Gene ID (Entrez) 9518 (Human); 23886 (Mouse); 29455 (Rat);
Precision
  • Intra-Assay Precision (Precision within an assay) Three samples of known concentration were tested on one plate to assess intra-assay precision
  • Inter-Assay Precision (Precision between assays) Three samples of known concentration were tested in separate assays to assess inter-assay precision
Quantity 1 Pack
Sample Volume Cell Culture Supernates (50uL),Serum (13uL),EDTA Plasma (13uL),Heparin Plasma (13uL),Urine (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym GDF15, growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1, MIC-1 NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1, NAG-1 NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDF, PDFGDF-15, PLAB, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGF-beta, PTGFBPTGF-beta
Total Assay Time 3.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.