Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human GDF-15 Quantikine QuicKit ELISA
SDP

Catalog No. QK957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QK957 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. QK957 Supplier R&D Systems™ Supplier No. QK957
Add to Cart
Add to Cart
This item is not returnable. View return policy

solid phase ELISA designed to measure human GDF-15 levels in cell culture supernates, serum, plasma, and urine

Specifications

Assay Range 0.28 to 1 pg/mL
Assay Sensitivity 0.947 pg/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum, Urine
Specificity Recognizes natural and recombinant human GDF-15
For Use With (Application) ELISA
Gene ID (Entrez) 9518
Quantity 1 Kit
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym GDF15, growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1, MIC-1 NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1, NAG-1 NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDF, PDFGDF-15, PLAB, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGF-beta, PTGFBPTGF-beta
Target Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.