Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GDF1 Full-length ORF (AAH22450.1, 1 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945690 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-690 10 ug
89-945-691 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-690 Supplier Abnova™ Supplier No. H00002657P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site that is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein is involved in the establishment of left-right asymmetry in early embryogenesis and in neural development in later embryogenesis. This protein is transcribed from a bicistronic mRNA that also encodes the longevity assurance gene. [provided by RefSeq]

Sequence: MAAAGPAAGPTGPEPMPSYAQLVQRGWGSALAAARGCTDCGWGLARRGLAEHAHLAPPELLLLALGALGWTALRSAATARLFRPLAKRCCLQPRDAAKMPESAWKFLFYLCSWSYSAYLLFGTDYPFFHDPPSVFYDWTPGMAVPRDIAAAYLLQGSFYGHSIYATLYMDTWRKDSVVMLLHHVVTLILIVSSYAFRYHNVGILVLFLHDISDVQLEFTKLNIYFKSRGGSYHRLHALAADLGCLSFGFSWFWFRLYWFPLKVLYATSHCSLRTVPDIPFYFFFNALLLLLTLMNLYWFLYIVAFAAKVLTGQVHELKDLREYDTAEAQSLKPSKAE

Specifications

Accession Number AAH22450.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2657
Molecular Weight (g/mol) 64.4kDa
Name GDF1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MAAAGPAAGPTGPEPMPSYAQLVQRGWGSALAAARGCTDCGWGLARRGLAEHAHLAPPELLLLALGALGWTALRSAATARLFRPLAKRCCLQPRDAAKMPESAWKFLFYLCSWSYSAYLLFGTDYPFFHDPPSVFYDWTPGMAVPRDIAAAYLLQGSFYGHSIYATLYMDTWRKDSVVMLLHHVVTLILIVSSYAFRYHNVGILVLFLHDISDVQLEFTKLNIYFKSRGGSYHRLHALAADLGCLSFGFSWFWFRLYWFPLKVLYATSHCSLRTVPDIPFYFFFNALLLLLTLMNLYWFLYIVAFAAKVLTGQVHELKDLREYDTAEAQSLKPSKAE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name GDF1
Gene Symbol GDF1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.