Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GDF3 Full-length ORF (NP_065685.1, 1 a.a. - 364 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945711 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-711 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-945-711 Supplier Abnova™ Supplier No. H00009573P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. [provided by RefSeq]

Sequence: MLRFLPDLAFSFLLILALGQAVQFQEYVFLQFLGLDKAPSPQKFQPVPYILKKIFQDREAAATTGVSRDLCYVKELGVRGNVLRFLPDQGFFLYPKKISQASSCLQKLLYFNLSAIKEREQLTLAQLGLDLGPNSYYNLGPELELALFLVQEPHVWGQTTPKPGKMFVLRSVPWPQGAVHFNLLDVAKDWNDNPRKNFGLFLEILVKEDRDSGVNFQPEDTCARLRCSLHASLLVVTLNPDQCHPSRKRRAAIPVPKLSCKNLCHRHQLFINFRDLGWHKWIIAPKGFMANYCHGECPFSLTISLNSSNYAFMQALMHAVDPEIPQAVCIPTKLSPISMLYQDNNDNVILRHYEDMVVDECGCG

Specifications

Accession Number NP_065685.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9573
Molecular Weight (g/mol) 67.8kDa
Name GDF3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLRFLPDLAFSFLLILALGQAVQFQEYVFLQFLGLDKAPSPQKFQPVPYILKKIFQDREAAATTGVSRDLCYVKELGVRGNVLRFLPDQGFFLYPKKISQASSCLQKLLYFNLSAIKEREQLTLAQLGLDLGPNSYYNLGPELELALFLVQEPHVWGQTTPKPGKMFVLRSVPWPQGAVHFNLLDVAKDWNDNPRKNFGLFLEILVKEDRDSGVNFQPEDTCARLRCSLHASLLVVTLNPDQCHPSRKRRAAIPVPKLSCKNLCHRHQLFINFRDLGWHKWIIAPKGFMANYCHGECPFSLTISLNSSNYAFMQALMHAVDPEIPQAVCIPTKLSPISMLYQDNNDNVILRHYEDMVVDECGCG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name GDF3
Gene Symbol GDF3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.